Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUZD1 Antibody (M4P6G6), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250186AF350
Description
CUZD1 Monoclonal specifically detects CUZD1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CUZD1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CUZD1 | |
Ovalbumin-conjugated synthetic peptide - RHFVNQRADY | |
0.1 mL | |
Primary | |
Human | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry | |
M4P6G6 | |
Western Blot, ELISA, Immunohistochemistry | |
CUB and zona pellucida-like domain-containing protein 1, CUB and zona pellucida-like domains 1, CUB and ZP domain-containing protein 1, ERG-1, estrogen regulated gene 1, Transmembrane protein UO-44, UO-44 | |
Mouse | |
Protein A purified | |
Cancer | |
50624.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction