Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CUZD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548129
Description
CUZD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
CUZD1 antiserum inhibits cell attachment and proliferation of ovarian cancer cells so may be involved in these processes. May also play a role in the uterus during late pregnancy and/or in trypsin activation in pancreatic acinar cells.
Specifications
CUZD1 | |
Polyclonal | |
Unconjugated | |
CUZD1 | |
CUB and zona pellucida like domains 1; CUB and zona pellucida-like domain-containing protein 1; CUB and zona pellucida-like domains 1; CUB and ZP domain-containing protein 1; CUZD1; ERG1; ERG-1; estrogen regulated gene 1; ITMAP1; Transmembrane protein UO-44; UNQ224/PRO257; UO-44 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
50624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86UP6 | |
CUZD1 | |
KLH conjugated synthetic peptide between 579-607 amino acids from the C-terminal region of human CUZD1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction