Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUZD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564378 Supplier Invitrogen™ Supplier No. PA564378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CKVLICDSSD HQSRCNQGCV SRSKRDISSY KWKTDSIIGP IRLKRDRSAS GNSGFQHETH AEETPNQP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

CUZD1 antiserum inhibits cell attachment and proliferation of ovarian cancer cells so may be involved in these processes. May also play a role in the uterus during late pregnancy and/or in trypsin activation in pancreatic acinar cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUZD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CUZD1
Gene Accession No. Q86UP6
Gene Alias CUB and zona pellucida like domains 1; CUB and zona pellucida-like domain-containing protein 1; CUB and zona pellucida-like domains 1; CUB and ZP domain-containing protein 1; CUZD1; ERG1; ERG-1; estrogen regulated gene 1; ITMAP1; Transmembrane protein UO-44; UNQ224/PRO257; UO-44
Gene Symbols CUZD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CUZD1. Recombinant protein control fragment (Product #RP-103684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.