Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CV 1808, Tocris Bioscience™

Catalog No. 171010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-101-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-101-0 Supplier R&D Systems Supplier No. 1710/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Non-selective adenosine A2 receptor agonist

Adenosine A2 receptor agonist. Coronary vasodilator, antihypertensive and antipsychotic following systemic administration in vivo.

Chemical Identifiers

CAS 53296-10-9
Molecular Formula C16H18N6O4
Molecular Weight (g/mol) 358.358
InChI Key SCNILGOVBBRMBK-SDBHATRESA-N
Synonym 2-phenylaminoadenosine, 2r,3r,4s,5r-2-6-amino-2-phenylamino-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, unii-k40i59g1ef, adenosine, 2-phenylamino, n6-phenylado, d06nrx, gtpl420, 3r,4s,5r-2-6-amino-2-anilino-purin-9-yl-5-hydroxymethyl oxolane-3,4-diol, 2-phenylamino ado cv-1808, 2-phenylaminoadenosine, solid
PubChem CID 6917803
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-anilinopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=CC=C(C=C1)NC2=NC3=C(C(=N2)N)N=CN3C4C(C(C(O4)CO)O)O

Specifications

Quantity 10 mg
Formula Weight 358.36
Percent Purity >99%
Chemical Name or Material CV 1808
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.