Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CWC15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244275
Description
Specifically detects CWC15 in Human samples, and it is validated for Immunoprecipitation
CWC15 Polyclonal specifically detects CWC15 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
CWC15 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
CWC15 | |
The epitope recognized by this antibody maps to a region between residue 75 to 125 of human Protein CWC15 homolog using the numbering given in entry Q9P013.2 (GeneID 51503). | |
100 μL | |
Primary | |
Human | |
IgG |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative) | |
C11orf5ORF5, CWC15 homolog, CWC15 homolog (S. cerevisiae), CWC15 spliceosome-associated protein homolog (S. cerevisiae), Cwf15, HSPC148, protein CWC15 homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
51503 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction