Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CWC15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23197725UL
Description
CWC15 Polyclonal specifically detects CWC15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CWC15 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| None | |
| CWC15 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RGGRGKGEGDLSQLSKQYSSRDLPSHTKIKYRQTTQDAPEEVRNRDFRRELEERERAAAREKNRDRPTREHTTSSSVS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C11orf5ORF5, CWC15 homolog, CWC15 homolog (S. cerevisiae), CWC15 spliceosome-associated protein homolog (S. cerevisiae), Cwf15, HSPC148, protein CWC15 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51503 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction