Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CWC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100462 Supplier Invitrogen™ Supplier No. PA5100462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CWC15.

Cwc15 gene ontology annotations related to this gene include mRNA splicing, via spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CWC15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cwc15
Gene Accession No. Q9JHS9, Q9P013
Gene Alias 0610040D20Rik; 2900052N06Rik; AD002; AD-002; C11orf5; CWC15; CWC15 homolog (S. cerevisiae); CWC15 spliceosome-associated protein; CWC15 spliceosome-associated protein homolog; CWC15 spliceosome-associated protein homolog (S. cerevisiae); Cwf15; Ed1; Embryonic development factor 1; HSPC148; mED1; nucleoporin; ORF5; protein CWC15 homolog; putative protein CWC15 like protein; spliceosome-associated protein CWC15 homolog
Gene Symbols Cwc15
Host Species Rabbit
Immunogen A synthesized peptide derived from human CWC15(Accession Q9P013), corresponding to amino acid residues M1-R44.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51503, 66070
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.