Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CWC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524276
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524276 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524276 Supplier Invitrogen™ Supplier No. PA524276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CWC22 Polyclonal Antibody for Western Blot, IHC (P), Flow

Required for pre-mRNA splicing as component of the spliceosome (PubMed:12226669, PubMed:11991638, PubMed:22961380, PubMed:28502770, PubMed:28076346, PubMed:29360106, PubMed:29301961). Promotes exon-junction complex (EJC) assembly (PubMed:22959432, PubMed:22961380). Hinders EIF4A3 from non-specifically binding RNA and escorts it to the splicing machinery to promote EJC assembly on mature mRNAs. Through its role in EJC assembly, required for nonsense-mediated mRNA decay. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CWC22
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CWC22
Gene Accession No. Q9HCG8
Gene Alias AA684037; AI173004; AL022752; B230213M24; Cwc22; CWC22 homolog, spliceosome-associated protein; CWC22 spliceosome-associated protein; CWC22 spliceosome-associated protein homolog; CWC22 spliceosome-associated protein homolog (S. cerevisiae); EIF4GL; fSAPb; functional spliceosome-associated protein b; KIAA1604; mKIAA1604; NCM; Nucampholin homolog; pre-mRNA-splicing factor CWC22 homolog; RGD1565385
Gene Symbols CWC22
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 16-45 amino acids from the N-terminal region of human CWC22.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57703
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.