Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CWC22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557796
Description
Immunogen sequence: RNQQTNDRKQ KERRQEHGHQ ETRTERERRS EKHRDQNSRG SNWRDPITKY TSDKDVPSER NNYSRVANDR DQEMHIDLEN KHGDP Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 48%.
Required for pre-mRNA splicing as component of the spliceosome (PubMed:12226669, PubMed:11991638, PubMed:22961380, PubMed:28502770, PubMed:28076346, PubMed:29360106, PubMed:29301961). Promotes exon-junction complex (EJC) assembly (PubMed:22959432, PubMed:22961380). Hinders EIF4A3 from non-specifically binding RNA and escorts it to the splicing machinery to promote EJC assembly on mature mRNAs. Through its role in EJC assembly, required for nonsense-mediated mRNA decay. [UniProt].
Specifications
CWC22 | |
Polyclonal | |
Unconjugated | |
CWC22 | |
AA684037; AI173004; AL022752; B230213M24; Cwc22; CWC22 homolog, spliceosome-associated protein; CWC22 spliceosome-associated protein; CWC22 spliceosome-associated protein homolog; CWC22 spliceosome-associated protein homolog (S. cerevisiae); EIF4GL; fSAPb; functional spliceosome-associated protein b; KIAA1604; mKIAA1604; NCM; Nucampholin homolog; pre-mRNA-splicing factor CWC22 homolog; RGD1565385 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCG8 | |
CWC22 | |
Recombinant protein corresponding to Human CWC22. Recombinant protein control fragment (Product #RP-96184). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction