Learn More
Invitrogen™ CWC22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588996
Description
Immunogen sequence: LTDELREHLK NTPKVIVAQK PDVEQNKSSP SSSSSASSSS ESDSSDSDSD SSDSSSESSS EESDSSSISS HSSASANDVR KKGHGKTRSK EVDKLIRNQQ TNDRKQKERR QEHGHQETRT ERERRSEKHR DQNSSGSNWR DPITKYTSDK DVPSERNNYS RVANDRDQEM HIDLENKHGD PKKKRGERRN SFSENEKHTH RIKDSENFRR KDRSKSKEMN RKHSGSRSDE DRYQNGAERR WEKSSRYSEQ SRESKKNQDR RREKSPAKQK; Positive Samples: Mouse liver, Mouse testis; Cellular Location: Nucleus, Nucleus speckle.
Required for pre-mRNA splicing as component of the spliceosome (PubMed:12226669, PubMed:11991638, PubMed:22961380, PubMed:28502770, PubMed:28076346, PubMed:29360106, PubMed:29301961). Promotes exon-junction complex (EJC) assembly (PubMed:22959432, PubMed:22961380). Hinders EIF4A3 from non-specifically binding RNA and escorts it to the splicing machinery to promote EJC assembly on mature mRNAs. Through its role in EJC assembly, required for nonsense-mediated mRNA decay. [UniProt].
Specifications
CWC22 | |
Polyclonal | |
Unconjugated | |
CWC22 | |
AA684037; AI173004; AL022752; B230213M24; Cwc22; CWC22 homolog, spliceosome-associated protein; CWC22 spliceosome-associated protein; CWC22 spliceosome-associated protein homolog; CWC22 spliceosome-associated protein homolog (S. cerevisiae); EIF4GL; fSAPb; functional spliceosome-associated protein b; KIAA1604; mKIAA1604; NCM; Nucampholin homolog; pre-mRNA-splicing factor CWC22 homolog; RGD1565385 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80744 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.04 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8C5N3 | |
CWC22 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 639-908 of human CWC22 (NP_0659941). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.