Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CWC25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545940 Supplier Invitrogen™ Supplier No. PA545940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RRETGQTRSP SPKKEVYQRR HAPGYTRKLS AEELERKRQE MMENAKWREE Sequence homology: Horse: 79%; Human: 100%; Rabbit: 86%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CWC25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CWC25
Gene Accession No. Q9NXE8
Gene Alias 1300013D05Rik; AA473636; CCDC49; coiled-coil domain containing 49; coiled-coil domain-containing protein 49; Cwc25; CWC25 spliceosome associated protein homolog; CWC25 spliceosome-associated protein homolog; CWC25 spliceosome-associated protein homolog (S. cerevisiae); pre-mRNA-splicing factor CWC25 homolog; R75228; Spliceosome-associated protein homolog CWC25
Gene Symbols CWC25
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CWC25 (aa 308-357).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54883
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.