Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CWC27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550750 Supplier Invitrogen™ Supplier No. PA550750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CWC27 protein.

As part of the spliceosome, plays a role in pre-mRNA splicing (PubMed:29360106). Probable inactive PPIase with no peptidyl-prolyl cis-trans isomerase activity (PubMed:20676357). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CWC27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Cwc27
Gene Accession No. Q6UX04
Gene Alias 3110009E13Rik; Antigen NY-CO-10; CWC27; CWC27 spliceosome associated protein homolog; CWC27 spliceosome-associated protein homolog; CWC27 spliceosome-associated protein homolog (S. cerevisiae); NY-CO-10; peptidyl-prolyl cis-trans isomerase CWC27 homolog; peptidyl-prolyl cis-trans isomerase SDCCAG10; PPIase CWC27; PPIase SDCCAG10; Probable inactive peptidyl-prolyl cis-trans isomerase CWC27 homolog; SDCCAG10; SDCCAG-10; Serologically defined colon cancer antigen 10; Spliceosome-associated protein CWC27 homolog; UNQ438/PRO871
Gene Symbols Cwc27
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human CWC27 spliceosome-associated protein homolog (S. cerevisiae).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10283
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.