Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CWC27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554993
Description
Immunogen sequence: EEEVKKLKPK GTKNFSLLSF GEEAEEEEEE VNRVSQSMKG KSKSSHDLLK DDPHLSSVPV VESEKGDAPD LVDDGEDESA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
As part of the spliceosome, plays a role in pre-mRNA splicing (PubMed:29360106). Probable inactive PPIase with no peptidyl-prolyl cis-trans isomerase activity (PubMed:20676357). [UniProt].
Specifications
CWC27 | |
Polyclonal | |
Unconjugated | |
Cwc27 | |
3110009E13Rik; Antigen NY-CO-10; CWC27; CWC27 spliceosome associated protein homolog; CWC27 spliceosome-associated protein homolog; CWC27 spliceosome-associated protein homolog (S. cerevisiae); NY-CO-10; peptidyl-prolyl cis-trans isomerase CWC27 homolog; peptidyl-prolyl cis-trans isomerase SDCCAG10; PPIase CWC27; PPIase SDCCAG10; Probable inactive peptidyl-prolyl cis-trans isomerase CWC27 homolog; SDCCAG10; SDCCAG-10; Serologically defined colon cancer antigen 10; Spliceosome-associated protein CWC27 homolog; UNQ438/PRO871 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UX04 | |
Cwc27 | |
Recombinant protein corresponding to Human CWC27. Recombinant protein control fragment (Product #RP-91751). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction