Learn More
CWC27 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | CWC27 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1015
![]() |
Bethyl Laboratories, Inc
A305685AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
PPIases accelerate the folding of proteins.Specifications
CWC27 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Peptidyl-prolyl cis-trans isomerase CWC27 homolog, PPIase CWC27, Antigen NY-CO-10, Serologically defined colon cancer antigen 10, CWC27, SDCCAG10 | |
Between 350 and 400 | |
Primary | |
4° C | |
CWC27 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q3TKY6, Q6UX04 | |
10283, 67285 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.