Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CWF19L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112995
Description
CWF19L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
CWF19L1 | |
Polyclonal | |
Unconjugated | |
CWF19L1 | |
2610528C06Rik; AI854304; AV336991; C19L1; Cwf19l1; CWF19-like 1 cell cycle control; CWF19-like 1, cell cycle control (S. pombe); CWF19-like protein 1; hDrn1; SCAR17 | |
Rabbit | |
Protein G | |
RUO | |
55280 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q69YN2 | |
CWF19L1 | |
Recombinant Human CWF19-like protein 1 protein (101-330AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction