Learn More
Description
CX-4945 can be used, in combination with other small molecules, in fast chemical reprogramming protocols for the induction of human somatic cells and highly efficient generation of pluripotent stem cells (hCiPS) in a minimum of 16 days with robust repeatability and reliability.
CX-4945 has antiproliferative effects in vitro as it attenuates PI3K/Akt signalling, causes cell-cycle arrest and selectively induces apoptosis in cancer cells. In models of angiogenesis, CX-4945 inhibits human umbilical vein endothelial cell migration, tube formation, and blocks CK2-dependent hypoxia-induced factor 1 alpha (HIF-1α) transcription in cancer cells.
In vivo, CX-4945 displays robust antitumor activity with concomitant reduction of the mechanism-based biomarker phospho-p21 (T145) (murine xenograft model). CX-4945 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | Sodium 5-[(3-Chlorophenyl)amino]benzo[c]-2,6-naphthyridine-8-carboxylate |
| CAS | 1309357-15-0 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Casein Kinase 2 Inhibitors |
| Molecular Formula | C19H11ClN3O2Na |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
