Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CX08005
GREENER_CHOICESDP

Catalog No. 61445 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-445 5 mg
61-442-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-445 Supplier Tocris Bioscience™ Supplier No. 6144/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Competitive PTP1B inhibitor

CX08005 is a competitive PTP1B inhibitor (IC50 = 0.781 μM). Binds catalytic P-loop of enzyme. Improves insulin sensitivity in obese mice in the glucose tolerance test and the homeostasis model assessment of insulin resistance index (HOMA-IR) . Enhances insulin-induced glucose uptake in adipocytes and myotubes in vitro. Exhibits some selectivity for PTP1B over other protein tyrosine phosphatases, except TCPTP.

Specifications

Chemical Name or Material 2-[[[2-(Tetradecyl)phenyl]amino]carbonyl]benzoic acid
CAS 1256341-22-6
Quantity 5 mg
Molecular Formula C28H39N04
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.