Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1 (Fractalkine) Mouse anti-Human, APC, Clone: V13-864, BD Pharmingen™

Catalog No. BDB567743 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567743 25 Tests
BDB567742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB567743 Supplier BD Biosciences Supplier No. 567743
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CX3CL1 (Fractalkine) Monoclonal antibody specifically detects CX3CL1 (Fractalkine) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CL1 (Fractalkine)
Applications Flow Cytometry
Classification Monoclonal
Clone V13-864
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias ABCD-3; C-X3-C motif chemokine 1; CXC3; CXC3C; NTN; NTT; SCYD1; neurotactin
Host Species Mouse
Immunogen Human CX3CL1 Recombinant Protein
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.