Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CX3CL1 Monoclonal Antibody (4C5)
Mouse Monoclonal Antibody
$222.50 - $569.00
Specifications
Antigen | CX3CL1 |
---|---|
Clone | 4C5 |
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
CX3CL1 Monoclonal Antibody for Western Blot, ELISA
Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.Specifications
CX3CL1 | |
1 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
O35188, P78423 | |
20312, 6376 | |
Non-glycosylated, polypeptide chain containing 76 amino acids of chemokine domain expressed in E. coli. | |
Antibody |
4C5 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
Cx3cl1 | |
A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1 | |
Cx3cl1 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title