Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1 Mouse anti-Human, Clone: 16A10, MilliporeSigma™

Catalog No. MABF229625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF229625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF229625 Supplier MilliporeSigma Supplier No. MABF229625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CX3CL1 Monoclonal antibody specifically detects CX3CL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 16A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen A recombinant fragment corresponding to 76 amino acids from the N-terminal, extracellular domain of human CX3CL1 (Fractalkine).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.