Learn More
CX3CL1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $2175.00
Specifications
Antigen | CX3CL1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P98BT25
![]() |
Invitrogen™
500P98BT25UG |
25 μg |
Each for $250.00
|
|
|||||
50-271-8427
![]() |
Invitrogen™
500P98BT500UG |
500 μg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Fractalkine (CX3CL1). Anti-Human Fractalkine (CX3CL1)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hFractalkine by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Fractalkine (500-P98) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hFractalkine. Western Blot: To detect hFractalkine by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hFractalkine is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P98BT-1MG will be provided as 2 x 500 μg
Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.Specifications
CX3CL1 | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Human | |
P78423 | |
6376 | |
E.coli-derived Recombinant Human Fractalkine (CX3CL1). | |
Antigen affinity chromatography | |
Cx3cl1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1 | |
Cx3cl1 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.