Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CR1 Rat anti-Human , Alexa Fluor 647, Clone: 2A9-1, BD Biosciences™

Catalog No. BDB565895 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565895 100 Tests
BDB565894 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565895 Supplier BD Biosciences Supplier No. 565895
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CR1
Applications Flow Cytometry
Classification Monoclonal
Clone 2A9-1
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor
Host Species Rat
Immunogen Human CX3CR1 Recombinant Protein
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.