Learn More
Description
Specifications
Specifications
| Antigen | CX3CR1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2A9-1 |
| Conjugate | Brilliant Blue 515 (BB515) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor |
| Host Species | Rat |
| Immunogen | Human CX3CR1 Recombinant Protein |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.