Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXADR Antibody (001), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290355PE
Description
CXADR Monoclonal antibody specifically detects CXADR in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CXADR | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
001 | |
ELISA, Immunohistochemistry-Paraffin | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
Rabbit immunized with purified, recombinant Mouse CXADR (Accession#: NP_001020363.1, Met1-Gly237) | |
0.1 mL | |
Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
1525 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction