Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXADR Antibody (020), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289642V
Description
CXADR Monoclonal antibody specifically detects CXADR in Human samples. It is validated for ELISASpecifications
CXADR | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
020 | |
ELISA | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR (NP_001329.1; Met1-Gly237). | |
0.1 mL | |
Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
1525 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction