Learn More
Invitrogen™ CXCL1/CXCL2/CXCL3 Monoclonal Antibody (29702)

Description
In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) CXCL2 is observed and no cross-reactivity with recombinant human (rh) VIC, rmVIC, rhCCL3L1, rhCCL4L1, or recombinant canine IL-8 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CXCL1/CXCL2/CXCL3 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 29702 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CXCL1 |
| Gene Accession No. | P09341, P19875, P19876 |
| Gene Alias | C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); chemokine (C-X-C motif) ligand 2; chemokine (C-X-C motif) ligand 3; CINC-2a; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine 2; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 1; C-X-C motif chemokine ligand 2; C-X-C motif chemokine ligand 3; CXCL; CXCL1; CXCL2; CXCL3; fibroblast secretory protein; FSP; GRO; GRO a; gro beta; gro gamma; GRO α; GRO1; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GRO2; GRO2 oncogene; GRO3; GRO3 oncogene; GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); GROB; Gro-beta; GRO-beta(5-73); GRO-beta-T; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated alpha protein; growth-regulated protein beta; Growth-regulated protein gamma; GROα; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2-alpha; macrophage inflammatory protein 2-beta; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; melanoma growth stimulatory activity beta; melanoma growth stimulatory activity gamma; MGSA; MGSA alpha; MGSA beta; MGSA gamma; MGSA-a; MGSA-b; MIP2; MIP2A; MIP-2a; MIP2-alpha; MIP2B; MIP-2b; MIP2-beta; NAP3; NAP-3; Neutrophil-activating protein 3; SB-251353; SCYB1; SCYB2; SCYB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.