Learn More
Invitrogen™ CXCL1 (GRO alpha) Monoclonal Antibody (KTYFLF), eFluor™ 660, eBioscience™

Description
Description: This KTYFLF monoclonal antibody reacts with human CXCL1 and has no cross-reactivity to human CXCL2 or CXCL3. CXCL1, or GRO (growth-related oncogene) alpha, is a pro-inflammatory CXC chemokine first identified by its constitutive overexpression in some tumors. It is closely related to CXCL2 (GRO beta) and CXCL3 (GRO gamma), with which it shares 90% and 86% sequence homology, respectively. These proteins, along with IL-8 or CXCL8, were later found to be critical for neutrophil mobilization and degranulation, as well as vascular permeabilization and angiogenesis. Signaling occurs through the G protein-coupled receptor CXCR2, which is shared with GRO beta and gamma. CXCR2 can also be activated by IL-8, although some studies suggest that the majority of IL-8 activity on neutrophils is mediated by CXCR1. CXCL1 is secreted by monocytes, epithelial cells, and fibroblasts in response to pro-inflammatory stimuli such as LPS, IL-1 beta, and TNF alpha. Overexpression is observed in many malignant tumors, where it contributes to tumor vascularization and metastasis. Applications Reported: This KTYFLF antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This KTYFLF antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of peripheral blood monocytes. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that wi...
Specifications
Specifications
| Antigen | CXCL1 (GRO alpha) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | KTYFLF |
| Concentration | 5 μL/Test |
| Conjugate | eFluor 660 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Cxcl1 |
| Gene Accession No. | P09341 |
| Gene Alias | alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51 |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.