Learn More
Invitrogen™ CXCL1 Monoclonal Antibody (48415)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA523745
Description
This antibody shows 100% cross-reactivity with recombinant human (rh) CXCL2, less than 40% cross-reactivity with recombinant rat (rr) CINC-1, and no cross-reactivity with rrCINC-3, rhCXCL3, rhCXCL5, rhCXCL6, or recombinant mouse (rm) CXCL6. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
Specifications
CXCL1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P12850 | |
Cxcl1 | |
E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1 Arg20-Lys96. | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Neutralization, Western Blot | |
48415 | |
Unconjugated | |
Cxcl1 | |
alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO a; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROa; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51 | |
Rat | |
Protein A/G | |
RUO | |
14825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.