Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1 Mouse anti-Human, Clone: 2A10, MilliporeSigma™

Catalog No. MABC1705100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1705100 100 μg
MABC170525 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1705100 Supplier MilliporeSigma Supplier No. MABC1705100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL1 Monoclonal antibody specifically detects CXCL1 in Human samples. It is validated for Western Blotting, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen Full-length mature recombinant human C-X-C motif chemokine 1 (CXCL1).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.