Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL10/IP-10/CRG-2 Antibody (6D4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259749MFV610
Description
CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Flow CytometrySpecifications
CXCL10/IP-10/CRG-2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
6D4 | |
Flow Cytometry | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
Recombinant human CXCL10/INP10 | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
3627 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction