Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cotton Rat CXCL10/IP-10/CRG-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1117SP
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Cotton Rat samples. It is validated for Western Blot, Neutralization.Specifications
CXCL10/IP-10/CRG-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 20-40 ug/mL | |
AAL16935 | |
CXCL10 | |
E. coli-derived recombinant cotton rat CXCL10/IP-10/CRG-2 Ile22-Val94 Accession # AAL16935 | |
25 μg | |
Primary | |
Detects cotton rat CXCL10/IP-10/CRG-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, or 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12, 13, 15 or 16, recombinant rat CXCL1, 2, 3, 5 or recombinant porcine CXCL8 is observed. | |
Cotton Rat | |
Purified |
Western Blot, Neutralization | |
163426 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3627 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction