Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCL10/IP-10/CRG-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | CXCL10/IP-10/CRG-2 |
---|---|
Clone | 1050502 |
Dilution | Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4661SP
![]() |
R&D Systems
MAB4661SP |
25 μg |
Each for $214.00
|
|
|||||
MAB4661100
![]() |
R&D Systems
MAB4661100 |
100 μg |
Each for $538.00
|
|
|||||
Description
CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Mouse samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
CXCL10/IP-10/CRG-2 | |
Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
E. coli-derived mouse CXCL10/IP-10/CRG-2 protein, Ile22-Pro98, Accession # Q548V9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1050502 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q548V9 | |
3627 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title