Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10 (IP10) Armenian Hamster anti-Mouse, Clone: 1B9, MilliporeSigma™

Catalog No. MABF234225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF234225 25 μL
MABF2342100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF234225 Supplier MilliporeSigma Supplier No. MABF234225UL
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CXCL10 (IP10) Monoclonal antibody specifically detects CXCL10 (IP10) in Mouse samples. It is validated for ELISA, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL10 (IP10)
Applications ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 1B9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified Amenian Hamster monoclonal antibody IgG1 in PBS without azide.
Host Species Armenian Hamster
Immunogen Full-length mature recombinant murine CXCL10 (IP10) produced in E. coli.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.