Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CXCL11 (Human) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Detection Limit:
<10 pg/mL
2 pg/mL
Specifications
Specifications
| Calibration Range | 62.5 to 4000 pg/mL |
| Content And Storage | Store the kit at -20°C |
| Description | CXCL11 (Human) is a sandwich enzyme immunoassay for the quantitative measurement of human CXCL11. |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Product Type | CXCL11 (Human) ELISA kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | 96 |
| For Use With (Application) | Quantification |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction