Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL11/I-TAC Antibody (10C6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222533H
Description
CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Human samples. It is validated for Western Blot, ELISA.Specifications
CXCL11/I-TAC | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CXCL11 | |
Full length recombinant protein expressed in E-Coli | |
0.2 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
6373 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
10C6 | |
Western Blot, ELISA | |
Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction