Learn More
Invitrogen™ CXCL11 Monoclonal Antibody (87328)
Description
In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL5, 7, 8, 9, or 10. In Western blots, this antibody shows less than 1% cross-reactivity with rhCXCL1, 2, 3, 5, 6, 7, IL-8/CXCL8, 9, 10, 13, CXCL12/SDF-1 alpha, CXCL12/SDF-1 beta, rmCXCL1, 2, 6, 9, 10, CXCL12/SDF-1 alpha, rmCXCL13, rrCXCL1, CXCL3/CINC-2 alpha, CXCL3/CINC-2 beta, or rpIL-8/CXCL8. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | CXCL11 |
| Applications | ELISA, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 87328 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CXCL11 |
| Gene Accession No. | O14625 |
| Gene Alias | betaR1; beta-R1; b-R1; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 11; CXC; CXC motif chemokine; C-X-C motif chemokine 11; C-X-C motif chemokine ligand 11; Cxc11; CXCL; CXCL11; H174; Interferon gamma-inducible protein 9; interferon-inducible T-cell alpha chemoattractant; IP9; IP-9; Itac; I-TAC; MGC102770; Scyb11; Scyb9b; small inducible cytokine B11; small inducible cytokine subfamily B (Cys-X-Cys), member 11; small inducible cytokine subfamily B (Cys-X-Cys), member 9B; small-inducible cytokine B11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.