Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CXCL11 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P132100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P132100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 500P132100 Supplier PeproTech Supplier No. 500P132100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human I-TAC (CXCL11). Anti-Human I-TAC (CXCL11)-specific antibody was purified by affinity chromatography employing an immobilized Human I-TAC (CXCL11) matrix. Sandwich ELISA: To detect Human I-TAC (CXCL11) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human I-TAC (CXCL11) (500-P132Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human I-TAC (CXCL11). Western Blot: To detect Human I-TAC (CXCL11) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human I-TAC (CXCL11) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Interferon-inducible T-cell alpha chemoattractant ( I-TAC), belonging to the CXC chemokine family, is expressed in peripheral blood leukocytes, pancreas and liver, thymus, spleen, lung, small intestine, placenta and prostate. Its molecular weight is 8-11 kD. I-TAC is also known as CXCL-11 and IP-9. The cytokine is involved in regulating calcium release and inducing a chemotactic response in activated T-cells and is the dominant ligand for CXC receptor-3. The gene encoding this protein contains 4 exons and at least three polyadenylation signals which might reflect cell-specific regulation of expression. IFN-gamma is a potent inducer of transcription of this gene. It is upregulated in patients with AIDS, dementia and in patients with multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CXCL11
Gene Accession No. O14625
Gene Alias betaR1; beta-R1; b-R1; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 11; CXC; CXC motif chemokine; C-X-C motif chemokine 11; C-X-C motif chemokine ligand 11; Cxc11; CXCL; CXCL11; H174; Interferon gamma-inducible protein 9; interferon-inducible T-cell alpha chemoattractant; IP9; IP-9; Itac; I-TAC; MGC102770; Scyb11; Scyb9b; small inducible cytokine B11; small inducible cytokine subfamily B (Cys-X-Cys), member 11; small inducible cytokine subfamily B (Cys-X-Cys), member 9B; small-inducible cytokine B11
Gene Symbols CXCL11
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human I-TAC (CXCL11).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6373
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.