Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL12/SDF-1 Antibody (MM0211-9N26), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212221MFV610
Description
CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human, Rat, Sheep samples. It is validated for Flow CytometrySpecifications
CXCL12/SDF-1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat, Sheep | |
Purified |
Flow Cytometry | |
MM0211-9N26 | |
Flow Cytometry | |
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
Human recombinant CXCL12/SDF-1 | |
0.1 mL | |
Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers | |
6387 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction