Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL12/SDF-1 beta Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305880MFV610
Description
CXCL12/SDF-1 beta Polyclonal antibody specifically detects CXCL12/SDF-1 beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CXCL12/SDF-1 beta | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
hSDF-1beta, SDF 1beta, SDF1B | |
Produced in rabbits immunized with purified, recombinant Human CXCL12/SDF-1 beta (Accession#: P48061-1; Lys22-Met93) | |
0.1 mL | |
Biologically Active Proteins | |
6387 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction