Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL13/BLC/BCA-1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216041G
Description
CXCL13/BLC/BCA-1 Polyclonal specifically detects CXCL13/BLC/BCA-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CXCL13/BLC/BCA-1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt′s lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
Rabbit | |
0.1 mL | |
10563 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50 mM sodium borate with 0.05% sodium azide | |
CXCL13 | |
Synthetic peptide made to a C-terminal region of human CXCL13 (between amino acids 50-109) [UniProt O43927] | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction