Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13/BLC/BCA-1 Antibody (MM0212-12A2) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50

Specifications

Antigen CXCL13/BLC/BCA-1
Clone MM0212-12A2
Concentration LYOPH
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019111
SDP
View Documents
Novus Biologicals
NBP2122220.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212222
SDP
View Documents
Novus Biologicals
NBP212222
0.1 mg N/A N/A N/A
Description

Description

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications

Specifications

CXCL13/BLC/BCA-1
LYOPH
Unconjugated
Mouse
Human
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
CXCL13
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
MM0212-12A2
Monoclonal
Purified
RUO
O43927
10563
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.