Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL13/BLC/BCA-1 Antibody (RM0205-19N1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2122230.025MG
Description
CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
CXCL13/BLC/BCA-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin, Blocking/Neutralizing 50-200 molar excess | |
O55038 | |
CXCL13 | |
Mouse CXCL13 | |
0.025 mg | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
Immunohistochemistry (Paraffin), Blocking Assay, Western Blot | |
RM0205-19N1 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
Rat | |
Protein G purified | |
RUO | |
10563 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction