Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13/BLC/BCA-1 Mouse anti-Human, Alexa Fluor 750, Clone: 53610, R&D Systems™

Catalog No. IC801S100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC801S100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC801S100UG Supplier R&D Systems Supplier No. IC801S100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL13/BLC/BCA-1
Applications Flow Cytometry
Classification Monoclonal
Clone 53610
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O43927
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Gene Symbols CXCL13
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL13/BLC/BCA-1 Val23-Arg94 Accession # O43927
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10563
Test Specificity Detects human CXCL13/BLC/BCA-1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL1, rhCXCL2, rhCXCL3, rhCXCL8/IL-8, rhCXCL10, rhCXCL9, rhCXCL12/SDF-1 alpha , or rhCXCL12/SDF-1 beta .
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.