Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL14/BRAK Antibody (MM0213-12B24), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212224AF532
Description
CXCL14/BRAK Monoclonal specifically detects CXCL14/BRAK in Human samples. It is validated for Western Blot.Specifications
CXCL14/BRAK | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CXCL14 | |
Human recombinant CXCL14/BRAK | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry | |
MM0213-12B24 | |
Western Blot, Immunohistochemistry | |
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14 | |
Mouse | |
Protein G purified | |
Biologically Active Proteins | |
9547.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction