Learn More
CXCL14/BRAK Antibody (RM0206-17P3), Alexa Fluor™ 750, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212225AF750
Catalog No. NP21222A70
Only null left
Add to Cart
Description
Description
CXCL14/BRAK Monoclonal specifically detects CXCL14/BRAK in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CXCL14/BRAK
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CXCL14
Mouse recombinant CXCL14/BRAK
0.1 mL
9547
Store at 4°C in the dark.
Western Blot
RM0206-17P3
Western Blot
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
Rat
Protein A or G purified
Primary
Mouse
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.