Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL14/BRAK Antibody (RM0206-17P3) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen CXCL14/BRAK
Clone RM0206-17P3
Concentration LYOPH
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019114
SDP
View Documents
Novus Biologicals
NBP2122250.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212225
SDP
View Documents
Novus Biologicals
NBP212225
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

CXCL14/BRAK Monoclonal specifically detects CXCL14/BRAK in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CXCL14/BRAK
LYOPH
Monoclonal
Purified
RUO
NP_062514
9547
IgG2
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
RM0206-17P3
Western Blot
Unconjugated
Rat
Mouse
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
CXCL14
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.