Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL14/BRAK Antibody (RM0206-17P3), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212225MFV610
Description
CXCL14/BRAK Monoclonal antibody specifically detects CXCL14/BRAK in Mouse samples. It is validated for Western BlotSpecifications
CXCL14/BRAK | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0206-17P3 | |
Western Blot | |
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14 | |
Mouse recombinant CXCL14/BRAK | |
0.1 mL | |
Biologically Active Proteins | |
9547 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction