Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL15/Lungkine Antibody (RM0207-16M7), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212416AF750
Description
CXCL15/Lungkine Monoclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.Specifications
CXCL15/Lungkine | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CXCL15 | |
Mouse recombinant CXCL15/Lungkine | |
0.1 mL | |
20309 | |
Store at 4°C in the dark. |
Western Blot | |
RM0207-16M7 | |
Western Blot | |
lungkine, Scyb15, weche | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction