Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL16 Antibody (4.4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | CXCL16 |
---|---|
Clone | 4.4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunofluorescence |
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CXCL16 | |
1 mg/mL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Mouse | |
Chemokines and Cytokines, Cytokine Research | |
PBS | |
58191 | |
IgG1 | |
Protein A purified |
4.4 | |
Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
GST-CXCL16 (143-224aa) fusion protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title