Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL16 Antibody (4.4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250187G
Description
CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CXCL16 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CXCL16 | |
GST-CXCL16 (143-224aa) fusion protein | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research | |
58191 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
4.4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction